Learn More
Analytical Sales & Services 10mm, 12 x 32 2mL Amber Wide Mouth Glass Screw Vials and 300µL Glass Bottom Spring Inserts - Pre-Assembled
Catalog No. 5000806028
Encompass
Advantage™ Pre-Assembled 10mm, 12 x 32 2mL Amber Wide Mouth Glass Screw Vials and 300µL Volume Glass Bottom Spring Inserts
Catalog No. 50-008-06028 Supplier Analytical Sales & Services Supplier No. 11255
Advantage™ Pre-Assembled 10mm, 12 x 32 2mL Amber Wide Mouth Glass Screw Vials and 300µL Volume Glass Bottom Spring Inserts