Learn More
Analytical Sales & Services 10mm, 12 x 32 2mL Amber Wide Mouth Screw Vials w/ Marking Spot and Caps w/ Pre-Slit Silicone/PTFE Liners
Catalog No. 5000805562
Encompass
Advantage™ Pre-Assembled Vial Kit\n\nContains: 10mm, 12 x 32 2mL Amber Glass Wide Mouth Screw Vials with Marking Spot and 10mm Black Screw Caps with Pre-Slit Silicone/PTFE Liners
Catalog No. 50-008-05562 Supplier Analytical Sales & Services Supplier No. 52348SLCASE
Advantage™ Pre-Assembled Vial Kit\n\nContains: 10mm, 12 x 32 2mL Amber Glass Wide Mouth Screw Vials with Marking Spot and 10mm Black Screw Caps with Pre-Slit Silicone/PTFE Liners