Learn More
Analytical Sales & Services 10mm, 12 x 32 2mL Amber Wide Mouth Screw Vials w/ Marking Spot and Caps w/ Pre-slit Silicone/PTFE Liners
Catalog No. 5000804750
Encompass
Advantage™ Unassembled Vial Kit\n\nContains: 10mm, 12 x 32 2mL Amber Glass Wide Mouth Screw Vials with Marking Spots and 10mm Black Screw Caps with Pre-slit Silicone/PTFE Liners
Catalog No. 50-008-04750 Supplier Analytical Sales & Services Supplier No. 53348ASLCASE
Add to Cart
Add to Cart
Advantage™ Unassembled Vial Kit\n\nContains: 10mm, 12 x 32 2mL Amber Glass Wide Mouth Screw Vials with Marking Spots and 10mm Black Screw Caps with Pre-slit Silicone/PTFE Liners