Learn More
Analytical Sales & Services 10mm, 12 x 32 2mL Amber Wide Mouth Screw Vials w/ Marking Spot and Caps w/ Silicone/PTFE Liners
Catalog No. 5000805071
Encompass
Advantage™ Unassembled Vial Kit. Contains: 10mm, 12 x 32 2mL Amber Glass Wide Mouth Screw Vials with Marking Spots and 10mm Black Screw Caps with Silicone/PTFE Liners
Catalog No. 50-008-05071 Supplier Analytical Sales & Services Supplier No. 53348ACASE
Advantage™ Unassembled Vial Kit. Contains: 10mm, 12 x 32 2mL Amber Glass Wide Mouth Screw Vials with Marking Spots and 10mm Black Screw Caps with Silicone/PTFE Liners