Learn More
Analytical Sales & Services 10mm, 12 x 32 2mL Clear Glass Wide Mouth Screw Vials with Marking Spots
Catalog No. 5000804827
Encompass
Advantage™ 10mm, 12 x 32 2mL Volume Clear Glass Wide Mouth Screw Vials with Marking Spots - for use with Shimadzu and Waters Autosamplers
Catalog No. 50-008-04827 Supplier Analytical Sales & Services Supplier No. 11231MCASE
Add to Cart
Add to Cart
Advantage™ 10mm, 12 x 32 2mL Volume Clear Glass Wide Mouth Screw Vials with Marking Spots - for use with Shimadzu and Waters Autosamplers