Learn More
Analytical Sales & Services 10mm, 12 x 32 2mL Clear Glass Wide Mouth Screw Vials - Silanized
Catalog No. 5000806564
Encompass
Advantage™ 10mm, 12 x 32 2mL Volume Clear Glass Wide Mouth Screw Vials - Silanized. For use with Shimadzu and Waters Autosamplers
Catalog No. 50-008-06564 Supplier Analytical Sales & Services Supplier No. 11231SCASE
Advantage™ 10mm, 12 x 32 2mL Volume Clear Glass Wide Mouth Screw Vials - Silanized. For use with Shimadzu and Waters Autosamplers