Learn More
Analytical Sales & Services 10mm, 12 x 32 2mL Clear Wide Mouth Screw Vials and Caps w/ Pre-Slit Silicone/PTFE Liners
Catalog No. 5000804979
Encompass
Advantage™ Pre-Assembled Vial Kit\n\nContains: 10mm, 12 x 32 2mL Clear Glass Wide Mouth Screw Vials and 10mm Black Screw Caps with Pre-Slit Silicone/PTFE Liners
Catalog No. 50-008-04979 Supplier Analytical Sales & Services Supplier No. 52009SLCASE
Advantage™ Pre-Assembled Vial Kit\n\nContains: 10mm, 12 x 32 2mL Clear Glass Wide Mouth Screw Vials and 10mm Black Screw Caps with Pre-Slit Silicone/PTFE Liners