Learn More
Analytical Sales & Services 10mm, 12 x 32 2mL Clear Wide Mouth Screw Vials and Caps w/ Silicone/PTFE Liners
Catalog No. 5000804983
Encompass
Advantage™ Unassembled Vial Kit. Contains: 10mm, 12 x 32 2mL Clear Glass Wide Mouth Screw Vials and 10mm Black Screw Caps with Silicone/PTFE Liners
Catalog No. 50-008-04983 Supplier Analytical Sales & Services Supplier No. 52248CASE
Advantage™ Unassembled Vial Kit. Contains: 10mm, 12 x 32 2mL Clear Glass Wide Mouth Screw Vials and 10mm Black Screw Caps with Silicone/PTFE Liners