Learn More
Analytical Sales & Services 10mm, 12 x 32 2mL Clear Wide Mouth Screw Vials w/ Marking Spot and Caps w/ Pre-Slit Silicone/PTFE Liners
Catalog No. 5000807072
Encompass
Advantage™ Unassembled Vial Kit. Contains: 10mm, 12 x 32 2mL Clear Glass Wide Mouth Screw Vials with Marking Spots and 10mm Black Screw Caps with Pre-Slit Silicone/PTFE Liners
Catalog No. 50-008-07072 Supplier Analytical Sales & Services Supplier No. 53348SLCASE
Advantage™ Unassembled Vial Kit. Contains: 10mm, 12 x 32 2mL Clear Glass Wide Mouth Screw Vials with Marking Spots and 10mm Black Screw Caps with Pre-Slit Silicone/PTFE Liners