Learn More
Analytical Sales & Services 10mm, 12 x 32 Clear Glass Wide Mouth Screw Thread MicroVials with 300µL Fused Glass Inserts - Silanized
Catalog No. 5000806950
Encompass
Advantage™ 10mm, 12 x 32 Clear Glass Wide Mouth Screw Thread MicroVials with 300µL Volume Fused Glass Inserts - Silanized
Catalog No. 50-008-06950 Supplier Analytical Sales & Services Supplier No. 11203S
Advantage™ 10mm, 12 x 32 Clear Glass Wide Mouth Screw Thread MicroVials with 300µL Volume Fused Glass Inserts - Silanized