Learn More
Ace Glass, Inc. 2mL 10mm Buchner filter funnel with porosity C (25-50 micron) ACE fritted glass disc.
SDP
Catalog No. 50979163
Encompass_Preferred
2mL 10mm Buchner filter funnel with porosity C (25-50 micron) ACE fritted glass disc.
Catalog No. 50-979-163 Supplier Ace Glass, Inc. Supplier No. 718606
Add to Cart
Add to Cart
2mL 10mm Buchner filter funnel with porosity C (25-50 micron) ACE fritted glass disc.