Learn More
Ace Glass, Inc. 2mL 10mm Buchner filter funnel with porosity D (10-20 micron) ACE fritted glass disc.
SDP
Catalog No. 50974271
Encompass_Preferred
2mL 10mm Buchner filter funnel with porosity D (10-20 micron) ACE fritted glass disc.
Catalog No. 50-974-271 Supplier Ace Glass, Inc. Supplier No. 718608
Add to Cart
Add to Cart
2mL 10mm Buchner filter funnel with porosity D (10-20 micron) ACE fritted glass disc.