Learn More
Ace Glass, Inc. 10mm 69cm solid glass stir shaft, polished, bead type
SDP
Catalog No. NC0320910
Encompass_Preferred
10mm O.D. polished glass stirring shaft; bead type. Length: 69cm. Use with only ACE PTFE stirrer bearings. Shafts accept 316 SS multi-paddle or turbine agitators secured with set-screw on ground flat section.
Catalog No. NC0320910 Supplier Ace Glass, Inc. Supplier No. 807536
10mm O.D. polished glass stirring shaft; bead type. Length: 69cm. Use with only ACE PTFE stirrer bearings. Shafts accept 316 SS multi-paddle or turbine agitators secured with set-screw on ground flat section.