Learn More
Acuderm Inc Acuderm Acu-Punch Biopsy Punch, Sterile, 10 mm, 50/bx
SDP
Catalog No. NC9236770
Encompass_Preferred
Biopsy Punch, Acu-Punch;, 10mm, Sterile, Only Disposable Skin Punch Made in the U.S.A, and Only Punch Available in a Complete Range of 14 Sizes, from 1 to 12mm, Always Razor Sharp and Ready to Use
Catalog No. NC9236770 Supplier Acuderm Inc Supplier No. P1050
Biopsy Punch, Acu-Punch;, 10mm, Sterile, Only Disposable Skin Punch Made in the U.S.A, and Only Punch Available in a Complete Range of 14 Sizes, from 1 to 12mm, Always Razor Sharp and Ready to Use