Learn More
Azzota Corp 10mm blk 0.1ml Z8.5 Quartz
Catalog No. 5000887167
Encompass_Preferred
Pathlength - 10mm Inside width - 2x5mm Volume - 0 1ml Dimensions - 45x12 5x12 5mm Black walls Z-Height - 8 5mm Type Q - Quartz (190nm-2500nm)
Catalog No. 50-008-87167 Supplier Azzota Corp Supplier No. Q6748.5MM
Pathlength - 10mm Inside width - 2x5mm Volume - 0 1ml Dimensions - 45x12 5x12 5mm Black walls Z-Height - 8 5mm Type Q - Quartz (190nm-2500nm)