Learn More
Azzota Corp 10mm blk 0.2ml Z15 Quartz
Catalog No. 5000887026
Encompass_Preferred
Pathlength - 10mm Inside width - 4x5mm Volume - 0 2ml Dimensions - 45x12 5x12 5mm Have black walls Z-Height - 15mm One year warranty Quartz (190nm-2500nm)
Catalog No. 50-008-87026 Supplier Azzota Corp Supplier No. Q676
Pathlength - 10mm Inside width - 4x5mm Volume - 0 2ml Dimensions - 45x12 5x12 5mm Have black walls Z-Height - 15mm One year warranty Quartz (190nm-2500nm)