Learn More
Azzota Corp 10mm cyl cuvette 2.8ml 1stop
Catalog No. 5000887065
Encompass_Preferred
10mm Pathlength Cylindrical Cuvettes 2 8ml Dimensions 22x12 5mm One stopper Quartz (190nm-2500nm)
Catalog No. 50-008-87065 Supplier Azzota Corp Supplier No. Q95
10mm Pathlength Cylindrical Cuvettes 2 8ml Dimensions 22x12 5mm One stopper Quartz (190nm-2500nm)