Learn More
Azzota Corp 10mm cyl cuvette 2.8ml 2stop
Catalog No. 5000887064
Encompass_Preferred
10mm Pathlength Cylindrical Cuvettes 2 8ml Dimensions 22x12 5mm Two stopper Quartz (190nm-2500nm)
Catalog No. 50-008-87064 Supplier Azzota Corp Supplier No. Q85
10mm Pathlength Cylindrical Cuvettes 2 8ml Dimensions 22x12 5mm Two stopper Quartz (190nm-2500nm)