Learn More
Across International 10mm Diameter ID Dry Pressing Die Set
SDP
Catalog No. 502251330
Encompass_Preferred
A set of 10mm diameter (ID) hardened steel dry pressing die set for pellet presses, ready for you to make 10mm diameter cylinder-shape pellets.
This item is not returnable. View return policy
Catalog No. 50-225-1330 Supplier Across International Supplier No. SDS10
A set of 10mm diameter (ID) hardened steel dry pressing die set for pellet presses, ready for you to make 10mm diameter cylinder-shape pellets.