Learn More
Azzota Corp 10mm flow cuvette 1.5mm 0.018m
Catalog No. 5000886975
Encompass_Preferred
10mm Pathlength Flowthrough Cuvettes Optical Glass 1 5mm Inside Width Volume 0 018ml
Catalog No. 50-008-86975 Supplier Azzota Corp Supplier No. G164
10mm Pathlength Flowthrough Cuvettes Optical Glass 1 5mm Inside Width Volume 0 018ml