Learn More
Azzota Corp 10mm Pathlength Standard Cuvette 3.5Ml - Pathlength 10mm - Inside Width 10mm Volume 3.5Ml Dimensions 45X12.5X12.5mm; Cover And Lid, Glass (340Nm-2500Nm)
Catalog No. 507532881
Encompass_Preferred
10mm Pathlength Standard Cuvette 3.5ml - Pathlength 10mm - Inside width 10mm Volume 3.5ml Dimensions 45x12.5x12.5mm; Cover and Lid, Glass (340nm-2500nm)
Catalog No. 50-753-2881 Supplier Azzota Corp Supplier No. G104
10mm Pathlength Standard Cuvette 3.5ml - Pathlength 10mm - Inside width 10mm Volume 3.5ml Dimensions 45x12.5x12.5mm; Cover and Lid, Glass (340nm-2500nm)