Learn More
Ace Glass, Inc. 10mm bushing, glass filled PTFE
SDP
Catalog No. 509558170
Encompass_Preferred
10mm glass-filled PTFE bushing, for use with self-aligning, lubricant free PTFE Trubore bearing or combination bearing / debris trap. Use with 10mm O.D. stainless steel or polished glass shafts.
Catalog No. 50-955-8170 Supplier Ace Glass, Inc. Supplier No. 806612
10mm glass-filled PTFE bushing, for use with self-aligning, lubricant free PTFE Trubore bearing or combination bearing / debris trap. Use with 10mm O.D. stainless steel or polished glass shafts.