Learn More
Ace Glass, Inc. 10MM GRADED SEALS 10MM GRADED SEALS
SDP
Supplier:  Ace Glass, Inc. 845516
Encompass
10mm quartz graded seals for connecting Pyrex or Kimax borosilicate glass to Quartz glass. Quartz length is 7.6cm, and borosilicate length is 10.2cm.
Catalog No. 50-979-781