Learn More
Azzota Corp 10mm Standard Ir Quartz Cuvette, 3.5Ml Pathlength: 10mm Volume/Capacity: 3.5Ml Wavelength Range: 250 To 3500Nm Exterior (Wlh) Is 12.5 X 12.5 X 45mm 2 Sides Of Polished Windows Glass
Catalog No. 507532890
Encompass_Preferred
10mm Standard IR Quartz Cuvettes - 3.5ml Pathlength: 10mm Volume/Capacity: 3.5 ml Material: IR Quartz Wavelength Range: 250 to 3500 nm Exterior (WLH) is 12.5 x 12.5 x 45 mm 2 sides of polished windows Glass/plastic lid and plastic box
Catalog No. 50-753-2890 Supplier Azzota Corp Supplier No. I104
10mm Standard IR Quartz Cuvettes - 3.5ml Pathlength: 10mm Volume/Capacity: 3.5 ml Material: IR Quartz Wavelength Range: 250 to 3500 nm Exterior (WLH) is 12.5 x 12.5 x 45 mm 2 sides of polished windows Glass/plastic lid and plastic box