Learn More
Chemglass Life Sciences 10mm Nylon Alignment Ring Only
SDP
Catalog No. 501531430
Encompass_Preferred
10mm Nylon Alignment Ring Only. Replacement part for the CG-2076 series of Chem-Stir stirrer bearings. Pictured is a fully assembled CHEM-STIR stirrer bearing. Item is Nylon Alignment Ring Only.
Catalog No. 50-153-1430 Supplier Chemglass Life Sciences Supplier No. CG207623
10mm Nylon Alignment Ring Only. Replacement part for the CG-2076 series of Chem-Stir stirrer bearings. Pictured is a fully assembled CHEM-STIR stirrer bearing. Item is Nylon Alignment Ring Only.