Learn More
Chemglass Life Sciences 10mm Diameter Quartz Rod, 48in Length
SDP
Catalog No. CGQ080109
Encompass_Preferred
10mm Diameter Quartz Rod, 48in Length. General Electric Type 214 clear fused quartz rod. Rod is stocked in 4ft lengths. PLEASE NOTE: PRICE LISTED IS PER LENGTH OF 4FT ROD.
Catalog No. CGQ080109 Supplier Chemglass Life Sciences Supplier No. CGQ080109
10mm Diameter Quartz Rod, 48in Length. General Electric Type 214 clear fused quartz rod. Rod is stocked in 4ft lengths. PLEASE NOTE: PRICE LISTED IS PER LENGTH OF 4FT ROD.