Learn More
Ace Glass, Inc. 10MM POR A TUBES (GB)
SDP
Supplier:  Ace Glass, Inc. 720502
Encompass_Preferred
10mm straight tubes with 145-174 micron fritted discs, for construction of special apparatus.
Catalog No. 50-979-197