Learn More
Ace Glass, Inc. 2mL 10mm Buchner filter funnel with porosity B (70-100 micron) ACE fritted glass disc.
SDP
Supplier:  Ace Glass, Inc. 718604
Encompass_Preferred
2mL 10mm Buchner filter funnel with porosity B (70-100 micron) ACE fritted glass disc.
Catalog No. 50-974-270