Learn More
Ace Glass, Inc. 10MM POR C TUBES (GB)
SDP
Catalog No. 50974374
Encompass_Preferred
10mm straight tubes with 25-50 micron fritted discs, for construction of special apparatus.
Catalog No. 50-974-374 Supplier Ace Glass, Inc. Supplier No. 720506
10mm straight tubes with 25-50 micron fritted discs, for construction of special apparatus.