Learn More
Ace Glass, Inc. 10MM POR D TUBES (GB)
SDP
Catalog No. 50974375
Encompass_Preferred
10mm straight tubes with 10-20 micron fritted discs, for construction of special apparatus.
Catalog No. 50-974-375 Supplier Ace Glass, Inc. Supplier No. 720508
10mm straight tubes with 10-20 micron fritted discs, for construction of special apparatus.