Learn More
Ace Glass, Inc. 10MM POR E TUBES (GB)
SDP
Catalog No. 50979198
Encompass_Preferred
10mm straight tubes with 4-8 micron fritted discs, for construction of special apparatus.
Catalog No. 50-979-198 Supplier Ace Glass, Inc. Supplier No. 720510
10mm straight tubes with 4-8 micron fritted discs, for construction of special apparatus.