Learn More
Ace Glass, Inc. 10mm PTFE inner bearing, debris trap style
SDP
Catalog No. 50975957
Encompass_Preferred
10mm self-aligning, lubricant-free PTFE Trubore bearing, debris trap style. Use with ACE polished stainless steel or glass 10mm O.D. stir shafts. Can achieve vacuum down to 1mm Hg. Multi-directional.
Catalog No. 50-975-957 Supplier Ace Glass, Inc. Supplier No. 806608
10mm self-aligning, lubricant-free PTFE Trubore bearing, debris trap style. Use with ACE polished stainless steel or glass 10mm O.D. stir shafts. Can achieve vacuum down to 1mm Hg. Multi-directional.