Learn More
Ace Glass, Inc. 10MM PTFE SADDLE O-RING 10MM PTFE SADDLE O-RING
SDP
Catalog No. 509558172
Encompass_Preferred
10mm compression shaft saddle with o-ring, for use with self-aligning, lubricant free PTFE Trubore stir shaft bearing assembly. Use with 10mm O.D. stainless steel or polished glass shafts.
Catalog No. 50-955-8172 Supplier Ace Glass, Inc. Supplier No. 806615
10mm compression shaft saddle with o-ring, for use with self-aligning, lubricant free PTFE Trubore stir shaft bearing assembly. Use with 10mm O.D. stainless steel or polished glass shafts.