Learn More
Buck Scientific Inc 10MM QUARTZ CUVETTE
SDP
Catalog No. NC9146190
Encompass
Type 1 10mm Quartz cuvette, Cell Type: 1, Path Length (mm): 10, Width (mm): 12.5, Height (mm): 45, Capacity (mL): 3.5
Catalog No. NC9146190 Supplier Buck Scientific Inc Supplier No. BS1Q10
Type 1 10mm Quartz cuvette, Cell Type: 1, Path Length (mm): 10, Width (mm): 12.5, Height (mm): 45, Capacity (mL): 3.5