Learn More
Azzota Corp 10mm Pathlength Standardir Cuvettes 3.5Ml - Pathlength 10mm Volume 3.5Ml Dimensions 12.5 X 12.5 X 45 mm; Glass/Plastic Lid And Plastic Box, Quartz (190nm To 2500 Nm)
Catalog No. 507532888
Encompass_Preferred
10mm Pathlength StandardIR Cuvettes 3.5ml - Pathlength 10mm Volume 3.5ml Dimensions 12.5 x 12.5 x 45 mm; Glass/plastic lid and plastic box, Quartz (250 to 3500 nm)
Catalog No. 50-753-2888 Supplier Azzota Corp Supplier No. Q104
10mm Pathlength StandardIR Cuvettes 3.5ml - Pathlength 10mm Volume 3.5ml Dimensions 12.5 x 12.5 x 45 mm; Glass/plastic lid and plastic box, Quartz (250 to 3500 nm)