Learn More
Chemglass Life Sciences 10mm Quartz Fritted Disc, Coarse
SDP
Catalog No. CGQ020702
Encompass_Preferred
10mm Quartz Fritted Disc, Coarse. Porous fused quartz discs. Porosity sizes are as listed and differ from porosities of borosilicate discs.
Catalog No. CGQ020702 Supplier Chemglass Life Sciences Supplier No. CGQ020702
10mm Quartz Fritted Disc, Coarse. Porous fused quartz discs. Porosity sizes are as listed and differ from porosities of borosilicate discs.