Learn More
Chemglass Life Sciences 10mm Quartz Fritted Disc, Medium
SDP
Catalog No. CGQ020701
Encompass_Preferred
10mm Quartz Fritted Disc, Medium. Porous fused quartz discs. Porosity sizes are as listed and differ from porosities of borosilicate discs.
Catalog No. CGQ020701 Supplier Chemglass Life Sciences Supplier No. CGQ020701
10mm Quartz Fritted Disc, Medium. Porous fused quartz discs. Porosity sizes are as listed and differ from porosities of borosilicate discs.