Learn More
Chemglass Life Sciences 10mm Quartz Fritted Disc, Extra Coarse
SDP
Catalog No. CGQ020703
Encompass_Preferred
10mm Quartz Fritted Disc, Extra Coarse. Porous fused quartz discs. Porosity sizes are as listed and differ from porosities of borosilicate discs.
Catalog No. CGQ020703 Supplier Chemglass Life Sciences Supplier No. CGQ020703
10mm Quartz Fritted Disc, Extra Coarse. Porous fused quartz discs. Porosity sizes are as listed and differ from porosities of borosilicate discs.