Learn More
Azzota Corp 10mm Quartz Std Cuvettes
Catalog No. 5000887062
Encompass_Preferred
10mm Pathlength Quartz Standard Cuvettes range 190-2500nm one stop
Catalog No. 50-008-87062 Supplier Azzota Corp Supplier No. Q14
10mm Pathlength Quartz Standard Cuvettes range 190-2500nm one stop