Learn More
Chemglass Life Sciences Chemglass CG-2077-M-210 10 mm Rotary Bearing Rebuild Kit for CG-2077-M Stirrer Bearings (with tools)
SDP
Catalog No. 502733536
Encompass_Preferred
Chemglass Life Sciences CG-2077-M-210 is a 10 mm rotary bearing rebuild kit designed for CG-2077-M series mechanical-seal stirrer bearings. The kit restores bearing seals and rolling elements and includes hand tools for disassembly and reassembly to support in-field maintenance of laboratory stirrers operating under vacuum or pressure.
  • Restores bearing seals and rolling elements.
  • Includes adjustable pin spanner wrench and retaining ring pliers.
  • Contains replacement ball bearings and needle roller bearings.
  • Designed for use with 10 mm diameter shafts.
  • Supports maintenance of vacuum and pressure integrity during operation.
  • Requires a small amount of specialized grease, sold separately.
Catalog No. 50-273-3536 Supplier Chemglass Life Sciences Supplier No. CG2077M210
Chemglass Life Sciences CG-2077-M-210 is a 10 mm rotary bearing rebuild kit designed for CG-2077-M series mechanical-seal stirrer bearings. The kit restores bearing seals and rolling elements and includes hand tools for disassembly and reassembly to support in-field maintenance of laboratory stirrers operating under vacuum or pressure.
  • Restores bearing seals and rolling elements.
  • Includes adjustable pin spanner wrench and retaining ring pliers.
  • Contains replacement ball bearings and needle roller bearings.
  • Designed for use with 10 mm diameter shafts.
  • Supports maintenance of vacuum and pressure integrity during operation.
  • Requires a small amount of specialized grease, sold separately.