Learn More
Chemglass Life Sciences 10mm Rotary Bearing Taper Lock, Ferrule, Nylon
SDP
Catalog No. 501531431
Encompass_Preferred
10mm Rotary Bearing Taper Lock, Ferrule, Nylon. Replacement part for the CG-2077-M series of mechanical seal stir bearings. Pictured is a fully assembled stir bearing. Item is ROTARY BEARING TAPER LOCK FERRULE ONLY.
Catalog No. 50-153-1431 Supplier Chemglass Life Sciences Supplier No. CG2077M110
10mm Rotary Bearing Taper Lock, Ferrule, Nylon. Replacement part for the CG-2077-M series of mechanical seal stir bearings. Pictured is a fully assembled stir bearing. Item is ROTARY BEARING TAPER LOCK FERRULE ONLY.