Learn More
Azzota Corp 10mm Pathlength (2X2.5mm Inside Width) Sub-Micro Quartz Cuvettes ..Volume - 0.05Ml, Z-Height 8.5mm
Catalog No. 501612921
Encompass_Preferred
10mm Pathlength (2x2.5mm Inside Width) Sub-micro Quartz Cuvettes ..Volume - 0.05ml, Z-height 8.5mm
Catalog No. 50-161-2921 Supplier Azzota Corp Supplier No. Q6758.5MM
10mm Pathlength (2x2.5mm Inside Width) Sub-micro Quartz Cuvettes ..Volume - 0.05ml, Z-height 8.5mm