Learn More
Azzota Corp 10mm semi-micro cuvette 1.75ml
Catalog No. 5000886974
Encompass_Preferred
10mm Pathlength (5mm Inside Width) Semi-micro Cuvettes Volume 1 75ml Optical Glass (340nm-2500nm)
Catalog No. 50-008-86974 Supplier Azzota Corp Supplier No. G144
10mm Pathlength (5mm Inside Width) Semi-micro Cuvettes Volume 1 75ml Optical Glass (340nm-2500nm)