Learn More
Chemglass Life Sciences 10mm Stirrer Shaft Retainer, Nylon, Safety
SDP
Catalog No. CG204801
Encompass_Preferred
10mm Safety Collar, Nylon. Designed to be placed on the stirrer shaft directly above the bearing. The collar provides protection against the stirrer shaft dropping and hitting the bottom of the vessel. The collar also acts as a dust cover and permits the stirrer to be positioned off the bottom of the vessel. Supplied complete with one each CG-2049 PTFE gasket of the corresponding size.
Catalog No. CG204801 Supplier Chemglass Life Sciences Supplier No. CG204801
10mm Safety Collar, Nylon. Designed to be placed on the stirrer shaft directly above the bearing. The collar provides protection against the stirrer shaft dropping and hitting the bottom of the vessel. The collar also acts as a dust cover and permits the stirrer to be positioned off the bottom of the vessel. Supplied complete with one each CG-2049 PTFE gasket of the corresponding size.