Learn More
Teknova 10mM Tris-HCl, pH 8.0, 20mL in 50mL Tube, 25 Tubes per Pack, DNase/RNase/Protease Tested, Sterile. 4 pack.
SDP
Catalog No. 502746004
Encompass
Tris (tris(hydroxymethyl)aminomethane) is used as a component ofbuffer solutions, especially for solutions ofnucleic acids. The useful buffer range for tris (pH 7–9) is similar to the pH of most living organisms. This, and its low cost, make tris one of the most common buffers in the biology/biochemistry laboratory.
Catalog No. 50-274-6004 Supplier Teknova Supplier No. T10204PK
Tris (tris(hydroxymethyl)aminomethane) is used as a component ofbuffer solutions, especially for solutions ofnucleic acids. The useful buffer range for tris (pH 7–9) is similar to the pH of most living organisms. This, and its low cost, make tris one of the most common buffers in the biology/biochemistry laboratory.