Learn More
Teknova 10mM Tris-HCl with 0.1% Tween-20, pH 8.5. 1mL/Tube, 50 Pack, Sterile.
SDP
Catalog No. NC0488943
Encompass
Tris (tris(hydroxymethyl)aminomethane) is used as a component ofbuffer solutions, especially for solutions ofnucleic acids. The useful buffer range for tris (pH 7–9) is similar to the pH of most living organisms. This, and its low cost, make tris one of the most common buffers in the biology/biochemistry laboratory.
This item is not returnable. View return policy
Catalog No. NC0488943 Supplier Teknova Supplier No. T7724
Tris (tris(hydroxymethyl)aminomethane) is used as a component ofbuffer solutions, especially for solutions ofnucleic acids. The useful buffer range for tris (pH 7–9) is similar to the pH of most living organisms. This, and its low cost, make tris one of the most common buffers in the biology/biochemistry laboratory.