Learn More
Ace Glass, Inc. 10mm x 46cm chromatography column
SDP
Supplier:  Ace Glass, Inc. 590219
Encompass_Preferred
10mm x 46cm chromatography column with #15 Ace-Thred at top and 2mm bore 1:5 taper solid PTFE stopcock plug at bottom. Column supplied with ACE 7506-06 nylon bushing and FETFE o-ring.
Catalog No. 50-978-592