Learn More
Ace Glass, Inc. 10mm x 46cm chromatography column
SDP
Catalog No. 50972570
Encompass_Preferred
10mm X 46cm chromatography column with 2mm bore, 1:5 taper solid PTFE stopcock plug, porosity B fritted disc sealed in at bottom, and a #15 Ace-Thred at top of column.
Catalog No. 50-972-570 Supplier Ace Glass, Inc. Supplier No. 590515
10mm X 46cm chromatography column with 2mm bore, 1:5 taper solid PTFE stopcock plug, porosity B fritted disc sealed in at bottom, and a #15 Ace-Thred at top of column.