Learn More
Abcam 10nm Gold Nanoparticles (1 OD), 20ML
Catalog No. 502662814
Encompass_Preferred
10nm Gold Nanoparticles (1 OD) (ab269933) is an aqueous suspension of spherical metallic nanoparticles manufactured to the highest standard to ensure: Uniform spherical shape, Ultra high quality, Batch to batch consistency and High efficiency antibody binding for maximum sensitivity.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-266-2814 Supplier Abcam Supplier No. AB26993320ML
10nm Gold Nanoparticles (1 OD) (ab269933) is an aqueous suspension of spherical metallic nanoparticles manufactured to the highest standard to ensure: Uniform spherical shape, Ultra high quality, Batch to batch consistency and High efficiency antibody binding for maximum sensitivity.
The product is subject to the following: Abcam Restricted Use Statement