Learn More
Abcam 10nm Gold Nanoparticles (10 OD), 20ML
Catalog No. 502656257
Encompass_Preferred
10nm Gold Nanoparticles (10 OD) (ab269934) is an aqueous suspension of spherical metallic nanoparticles.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-265-6257 Supplier Abcam Supplier No. AB26993420ML
10nm Gold Nanoparticles (10 OD) (ab269934) is an aqueous suspension of spherical metallic nanoparticles.
The product is subject to the following: Abcam Restricted Use Statement